Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
SDP
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
$150.15 - $159.60
Specifications
For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm
View More Specs
Products 14
Catalog Number Mfr. No. Thickness (English) Thickness (Metric) Volume (Metric) Well No. of Wells Price Quantity & Availability  
Catalog Number Mfr. No. Thickness (English) Thickness (Metric) Volume (Metric) Well No. of Wells Price Quantity & Availability  
03-500-173
SDP
View Documents
Hoefer™
SE211A91.0
0.03 in. 0.1 cm 5.8μL/ mm 9
Each for $150.15
Add to cart
03-500-169
SDP
Accessories View Documents
Hoefer™
SE211A181.0
0.03 in. 0.1 cm 2.9μL/ mm 18
Each for $150.15
Add to cart
03-500-176
SDP
Accessories View Documents
Hoefer™
SE211AR1.5
0.05 in. 0.15 cm 120μL/ mm 1 Prep, 1 Reference
Each for $154.35
Add to cart
03-500-166
SDP
Accessories View Documents
Hoefer™
SE211A15.75
0.02 in. 0.07 cm 2.2μL/ mm 15
Each for $154.35
Add to cart
03-500-174
SDP
Accessories View Documents
Hoefer™
SE211AR.75
0.02 in. 0.07 cm 51μL/ mm 1 Prep, 1 Reference
Each for $150.15
Add to cart
03-500-170
SDP
Accessories View Documents
Hoefer™
SE211A5.75
0.02 in. 0.07 cm 9.8μL/ mm 5
Each for $151.20
Add to cart
03-500-163
SDP
Accessories View Documents
Hoefer™
SE211A10.75
0.03 in. 0.075 cm 9.8μL/ mm 10
Each for $154.35
Add to cart
03-500-168
SDP
Accessories View Documents
Hoefer™
SE211A151.5
0.05 in. 0.15 cm 4.4μL/ mm 15
Each for $154.35
Add to cart
03-500-167
SDP
Accessories View Documents
Hoefer™
SE211A151.0
0.03 in. 0.1 cm 2.9μL/ mm 15
Each for $154.35
Add to cart
03-500-165
SDP
Accessories View Documents
Hoefer™
SE211A101.5
0.05 in. 0.15 cm 7.2μL/ mm 10
Each for $154.35
Add to cart
03-500-172
SDP
Accessories View Documents
Hoefer™
SE211A51.5
0.05 in. 0.15 cm 19.5μL/ mm 5
Each for $150.15
Add to cart
03-500-164
SDP
Accessories View Documents
Hoefer™
SE211A101.0
0.03 in. 0.1 cm 4.8μL/ mm 10
Each for $154.35
Add to cart
03-500-171
SDP
Accessories View Documents
Hoefer™
SE211A51.0
0.03 in. 0.1 cm 13μL/ mm 5
Each for $159.60
Add to cart
03-500-175
SDP
Accessories View Documents
Hoefer™
SE211AR1.0
0.03 in. 0.1 cm 68μL/ mm 1 Prep, 1 Reference
Each for $154.35
Add to cart
Description
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
  • Lightweight
  • Virtually unbreakable
Specifications
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
PTFE
1.3 cm
Comb
0.51 in.