Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units Available on GSA/VA Contract for Federal Government customers only.

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Manufacturer: Hoefer™ SE51120.75

 View more versions of this product

Catalog No. 03-500-191

  Check Availability
Add to Cart

    Specifications

    Width (Metric) 0.41cm
    Depth (English) 0.98 in.
    Depth (Metric) 2.5cm
    For Use With (Equipment) SE 600, SE 400 series vertical electrophoresis system
    Product Type Comb
    No. of Wells 20
    Thickness (English) 0.02 in.
    Volume (Metric) Well 3.1μL/mm
    Material PTFE
    Thickness (Metric) 0.07cm

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit