Hoefer™ Gel Casters for Mini Vertical Electrophoresis Systems Available on GSA/VA Contract for Federal Government customers only.
Easy-to-use casters
Manufacturer: Hoefer™ SE275
 View more versions of this product
Catalog No. 03-500-491
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) SE 250 mini vertical gel electrophoresis unit
    Product Type Gel Caster
    Includes Casting cradle with gasket, face, Glass, alumina, space saver plates, filler sheets, plugs
    No. of Gels 1 to 4
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback