Hoefer™ HE 99X Max Submarine Electrophoresis Unit Available on GSA/VA Contract for Federal Government customers only.

Offers no-mess gel casting without tape or dams

Manufacturer: Hoefer™ HE99X151.5

 View more versions of this product

Catalog No. 03-500-121

  Check Availability
Add to Cart

    Specifications

    Width (English) Exterior 7.1 in.
    Length (English) Gel 5.9, 3.9 or 7.8 in.
    Temperature (Metric) Maximum 45°C
    Length (English) Exterior 14.1 in.
    Width (Metric) Exterior 18.2cm
    Volume (English) Buffer 40.5 oz.
    Volume (Metric) Buffer 1200mL
    Width (English) Gel 5.9 in.
    Height (English) Exterior 5.5 in.
    Sample Size 60
    Product Type Horizontal Agarose electrophoresis unit
    Height (Metric) Exterior 14cm
    Length (Metric) Exterior 36cm
    Certifications/Compliance EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
    Length (Metric) Gel 15, 10 or 20cm
    Width (Metric) Gel 15cm
    Amperage 100mA
    Dimensions (L x W x H) Exterior 14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14cm)
    Humidity Range Up to 80%
    Includes Buffer chamber assembly, safety lid with high voltage Leads, 15 x 20cm running tray, 15 x 20cm casting tray, Foam gaskets, bubble level, 1.5mm thick, 15-well comb, Comb Back
    Temperature (English) Maximum 113°F
    Voltage 200V
    Wattage 20w

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit