Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hoefer HE 99X Max Submarine Electrophoresis Unit
Offers no-mess gel casting without tape or dams
$670.00 - $826.00
Specifications
Amperage | 100 mA |
---|---|
Certifications/Compliance | EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE |
Dimensions (L x W x H) | 14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm) |
Length (English) Gel | 5.9, 3.9 or 7.8 in. |
Length (Metric) Gel | 15, 10 or 20 cm |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
03-500-120
|
Hoefer
HE99X |
Buffer chamber assembly, safety lid with high voltage Leads, Foam gaskets, bubble level |
Each for $670.00
|
|
03-500-121
|
Hoefer
HE99X151.5 |
Buffer chamber assembly, safety lid with high voltage Leads, 15 x 20cm running tray, 15 x 20cm casting tray, Foam gaskets, bubble level, 1.5mm thick, 15-well comb, Comb Back |
Each for $826.00
|
|
Description
- Unit accommodates a variety of gel lengths for maximum resolution of DNA fragments
- Agarose bridges are also accommodated, so gels can be run in nonsubmarine fashion
- Rugged design provides reliability and safety
Specifications
100 mA | |
14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm) | |
15, 10 or 20 cm | |
40.5 oz. | |
5.9 in. | |
Up to 80% | |
113°F | |
200 V |
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
5.9, 3.9 or 7.8 in. | |
Horizontal Agarose Electrophoresis Unit | |
1200 mL | |
15 cm | |
60 | |
45°C | |
20 w |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title