Learn More
Hoefer™ HE 99X Max Submarine Electrophoresis Unit
Offers no-mess gel casting without tape or dams
Manufacturer:  Hoefer™ HE99X
 View more versions of this product
Catalog No. 03-500-120
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
100mA
Up to 80%
60
45°C
20w
40.5 oz.
5.9, 3.9 or 7.8 in.
5.9 in.
5.5 in.
14.1 in.
7.1 in.
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14cm)
Buffer chamber assembly, safety lid with high voltage Leads, 15 x 20cm running tray, 15 x 20cm casting tray, Foam gaskets, bubble level
113°F
200V
Horizontal Agarose electrophoresis unit
1200mL
15, 10 or 20cm
15cm
14cm
36cm
18.2cm
Accessories