Hoefer™ HE-PLUS Horizontal Electrophoresis System

Compact, easy-to-use system completely integrated with a built-in variable power supply

Manufacturer: Hoefer™ HEPLUS115V

 View more versions of this product

Catalog No. 03-500-553

  Check Availability
Add to Cart

    Specifications

    Height (Metric) Exterior 6.2cm
    Length (English) Exterior 6.6 in.
    Volume (English) Buffer 11.8 oz.
    Length (English) Gel 5.1 in.
    Width (English) Gel 4.9 in.
    Dimensions (L x W x H) Exterior 6.6 x 9.6 x 2.4 in. (17 x 24.5 x 6.2cm)
    Display Type LED
    Electrical Requirements 115V 50/60Hz
    Hertz 50/60Hz
    Includes Gel tank, safety lid, power supply, large gel tray, medium gel trays, large or medium comb 14, 28-well, casting stand
    Sample Size 112
    Timer 99.59 Hours or continuous run
    Voltage 115V
    Wattage 45w
    Weight (English) 0.9 lb.
    Product Type Horizontal electrophoresis system
    Length (Metric) Exterior 17cm
    Width (English) Exterior 9.6 in.
    Width (Metric) Exterior 24.5cm
    Certifications/Compliance CE, CSA, TVE
    Volume (Metric) Buffer 350mL
    Length (Metric) Gel 13cm
    Width (Metric) Gel 12.5cm
    Height (English) Exterior 2.4 in.
    Weight (Metric) 410g

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit