Learn More
Hoefer™ Maxi-Plus and Maxi-Plus Submarine Gel Electrophoresis Units
Ideal for high resolution analytical and preparative studies of nucleic acids
Manufacturer:  Hoefer™ SUB25
 View more versions of this product
Catalog No. 03-500-792
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
500mA
High throughput screening of Nucleic Acids, particularly PCR products and in genotyping studies
80%
22.04 in.
624
45°C
500w
9.8 in.
11.8 in.
EN61010-1, CE
22.04 x 13.77 in. (56 x 35cm)
25 x 30cm
Buffer chamber assembly, safety lid with high voltage Leads, running tray with casting gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
56cm
113°F
1000V
Maxi plus standard submarine gel electrophoresis unit
25cm
30cm