Hoefer™ Maxi-Plus and Maxi-Plus Submarine Gel Electrophoresis Units

Ideal for high resolution analytical and preparative studies of nucleic acids

Manufacturer: Hoefer™ SUB20C

 View more versions of this product

Catalog No. 03-500-788

  Check Availability
Add to Cart

    Specifications

    Height (Metric) Exterior 8cm
    Length (English) Exterior 18.7 in.
    Width (English) Exterior 10.6 in.
    Certifications/Compliance EN61010-1, CE
    Volume (Metric) Buffer 2200mL
    Length (Metric) Gel 20cm
    Width (English) Gel 7.8 in.
    Temperature (English) Maximum 113°F
    Wattage 500w
    Length (Metric) Exterior 47.5cm
    Width (Metric) Exterior 27cm
    Volume (English) Buffer 74.3 oz.
    Length (English) Gel 7.8 in.
    Width (Metric) Gel 20cm
    Height (English) Exterior 3.1 in.
    Amperage 500mA
    Dimensions (L x W x H) Exterior 18.7 x 10.6 x 3.1 in. (47.5 x 27 x 8cm)
    Humidity Range Up to 80%
    Includes Buffer chamber assembly with cooling ports, safety lid with high voltage Leads, running tray with casting gates, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
    Sample Size 160
    Temperature (Metric) Maximum 45°C
    Voltage 1000V
    Product Type Submarine gel electrophoresis unit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit