Hoefer™ Mini-Peristaltic Pump

For use with horizontal units with buffer recirculation ports

Manufacturer: Hoefer™ PP24

 View all options for this product

Catalog No. 03-500-411


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Height (Metric) 189cm
    Length (English) 41.3 in.
    Length (Metric) 105cm
    Width (English) 44.88 in.
    Width (Metric) 114cm
    Amperage 800mA
    Certifications/Compliance EN61010-1, CE
    Product Type Mini Peristaltic Pump
    For Use With (Equipment) Horizontal unit with Buffer recirculation port
    Voltage 115/230V
    Wattage 10w
    Dimensions (L x W) 41.3 x 44.88 in. (105 x 114cm)

    The Mini-Peristaltic Pump can be used with either a single tube or two tubes simultaneously. Two of the Pump Head Tubing Pieces are included with the pump. Additional Pump Head Tubing Pieces may be purchased separately.

    • Two front panel controls provide flow rates from approximately 0.8 to 24.5mL/min.
    • Control knob provides variable adjustment from 0 to 100% of the selected flow rate range
    • Second control is a two position toggle switch marked x1, x2 which selects low or high flow rates
    • Four roller pump head is on top of the stout metal box
    • Back of the pump head rotates into an“open”position and either one or two tubes can be dropped into slots
    • Loaded section rotates back against spring loaded jaws and locks into place
    • Tubing is automatically in proper wiping contact with the pump head rollers
    • Each pump is provided with a 12.5mm (0.5 in.) rod clamp on the back so that multiple pumps can be mounted vertically on a lattice rod

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit