Learn More
Hoefer™ Mini and Standard 2-D Electrophoresis Systems
Offer a versatile, powerful focusing unit for the first dimension and an easy-to-use, highly reproducible, second-dimension separation tank
Manufacturer:  Hoefer™ 526003
 View more versions of this product
Catalog No. 52-600-3
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
500mA
IEF100 unit, SE 260-10A-.75 Mighty Small™ II mini deluxe vertical unit, 10-well 0.75mm combs, spacers, multiple gel caster with accessories
300V
0.33 oz.
4.13 in.
10cm
0.29 in.
Mini 2-D Combination System
90w
10mL
3.93 in.
10.5cm