Hoefer™ Mini and Standard 2-D Electrophoresis Systems

Offer a versatile, powerful focusing unit for the first dimension and an easy-to-use, highly reproducible, second-dimension separation tank

Manufacturer: Hoefer™ 560003

 View more versions of this product

Catalog No. 56-000-3

  Check Availability
Add to Cart

    Specifications

    Product Type Standard 2-D Combination System
    Length (Metric) Exterior 16cm
    Volume (English) Buffer 1.69 oz.
    Volume (Metric) Buffer 50mL
    Length (English) Gel 6.29 in.
    Width (English) Gel 7.08 in.
    Width (Metric) Gel 18cm
    Amperage 500mA
    Height (English) Gel 0.59 in.
    Includes IEF100 unit deluxe dual cooled vertical electrophoresis unit and PS00B, multiple gel caster with accessories
    Voltage 300V
    Wattage 90w

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit