Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Economical equipment packages provide proven electrophoresis solutions for Western blotting
Manufacturer: Hoefer™ 326223
 View more versions of this product
Catalog No. 03-500-804
  Check Availability
Add to Cart
    Specifications
    Product Type Mini Electrophoresis Blotting System
    Includes Mighty Small™ II mini deluxe vertical unit, Mighty Small™ mini transfer tank, power supply
    Amperage 500mA
    Voltage 300V
    Wattage 90w
  • Description & Specifications
Related Products
Product Content Feedback