Hoefer™ Replacement Parts for HE 99X Max Submarine Electrophoresis Unit Available on GSA/VA Contract for Federal Government customers only.
Various replacement parts for Submarine Electrophoresis Unit
Manufacturer: Hoefer™ HE90X
 View more versions of this product
Catalog No. 03-500-432
  Check Availability
Add to Cart
    Specifications
    Product Type Buffer Chamber Assembly
    For Use With (Equipment) HE99X max submarine electrophoresis unit
    Includes 4 Leveling feet, 2 banana plugs with Nickel and Nylon washers and 2 Platinum wire electrode strip assemblies
  • Description & Specifications
Related Products
Product Content Feedback