Hoefer™ SE 400 Series Sturdier™ Air-Cooled Vertical Electrophoresis Systems Available on GSA/VA Contract for Federal Government customers only.

Economical units ideal for applications in which temperature control is not crucial

Manufacturer: Hoefer™ SE400151.5

 View more versions of this product

Catalog No. 03-500-500

  Check Availability
Add to Cart

Required Accessories : Comb and spacers must be ordered separately for Model SE 410 Basic.

    Specifications

    Thickness (Metric) Spacer 0.15cm
    Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
    Volume (English) Buffer 23.66 oz.
    Width (English) Gel 7.08 in.
    Depth (English) 5.9 in.
    Includes Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal, comb, 15-well, 1.5 mm thick, spacers
    Temperature (English) Maximum 113°F
    Width (English) Exterior 9.44 in.
    Width (Metric) Exterior 24cm
    Volume (Metric) Buffer 700mL
    Width (Metric) Gel 18cm
    Height (English) Exterior 11.02 in.
    Height (Metric) Exterior 28cm
    Amperage 60mA
    Depth (Metric) 15cm
    Height (English) Gel 6.29 in.
    Humidity Range Up to 80%
    Temperature (Metric) Maximum 45°C
    Temperature (Metric) Storage 15° to 30°C
    Thickness (English) Spacer 0.05 in.
    Voltage 500V
    Wattage 20w

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit