Hoefer™ SG Series Gradient Makers Available on GSA/VA Contract for Federal Government customers only.
Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride
Manufacturer: Hoefer™ SG30
 View more versions of this product
Catalog No. 03-500-277
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Mighty Small™ gel caster SE 275 (for 4 gels) or SE 215 (for 10 gels)
    Product Type Gradient Maker
    Includes Gradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle
    Volume (English) 1 oz.
    Volume (Metric) 30mL
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback