Learn More
Hoefer™ SG Series Gradient Makers
Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride
Manufacturer:  Hoefer™ SG50
 View more versions of this product
Catalog No. 03-500-278
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Mighty Small™ gel caster SE 275 (for 4 gels) or SE 215 (for 10 gels)
Gradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle
50mL
Gradient Maker
1.69 oz.
Accessories