Hoefer™ SG Series Gradient Makers Available on GSA/VA Contract for Federal Government customers only.
Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride
Manufacturer: Hoefer™ SG500
 View more versions of this product
Catalog No. 03-500-279
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) SE 615, SE 915 Multiple gel casters
    Product Type Gradient Maker
    Includes Gradient maker w/Push-Pull Valve, four wrap-and-seal hose clamps
    Volume (English) 16.9 oz.
    Volume (Metric) 500mL
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback