Learn More
Hoefer™ SUB10 Mini-Plus Submarine Combs
Accessories for SUB10 Mini-Plus Submarine
Manufacturer:  Hoefer™ SUB10C116
 View more versions of this product
Catalog No. 03-500-724
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.04 in.
0.19 in.
Comb
16
15μL
0.1cm
0.5cm
SUB10 mini plus submarine unit
0.14 in.
0.36cm