Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Sponges Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Hoefer™ TE45
 View more versions of this product
Catalog No. 03-500-292
  Check Availability
Add to Cart
    Specifications
    Material Dacron
    Thickness (English) 0.25 in.
    Thickness (Metric) 0.6cm
    Quantity 4
    For Use With (Equipment) TE 42, TE 62 transfer tank
    Product Type Sponge
  • Description & Specifications
Related Products
Product Content Feedback