Hoefer™ TE42 and TE62 Tank Blotting Units Available on GSA/VA Contract for Federal Government customers only.

Perform high-intensity protein transfers in one hour

Manufacturer: Hoefer™ TE42

 View more versions of this product

Catalog No. 03-500-217

  Check Availability
Add to Cart

    Specifications

    Product Type Standard Transfer Tank
    Amperage 2A
    Temperature (Metric) Maximum 45°C
    Wattage 200w
    Height (Metric) Exterior 13cm
    Length (Metric) Exterior 30.5cm
    Width (Metric) Exterior 28cm
    Dimensions (L x W x H) Exterior 12 x 11.02 x 5.11 in. (30.5 x 28 x 13cm)
    Includes Lower chamber, safety lid with high voltage Leads, 2 electrode panels, 2 gel cassettes, four 0.13 in. thick Foam sponges, two 0.25 in. thick Foam sponges, 25 sheets blotting Paper, cassette hook
    Volume (English) Buffer 135.25 to 169.07 oz.
    Device Holds Up to two 15 x 21cm gel (with cooling), up to four 15 x 21cm gel (without cooling)
    Humidity Range Up to 80%
    Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
    Temperature (English) Maximum 113°F
    Volume (Metric) Buffer 4000 to 5000mL
    Voltage 100V
    Height (English) Exterior 5.11 in.
    Length (English) Exterior 12 in.
    Width (English) Exterior 11.02 in.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit