Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ TE70X and TE77X Semidry Blotting Systems
SDP

Perform uniform transfers of proteins and nucleic acids from polyacrylamide and agarose gels in less than one hour

$2130.00 - $2750.00

Specifications

Amperage 500 mA
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Dimensions (L x W x H) 18.89 x 15 x 4 in. (48 x 38 x 10 cm)
For Use With (Equipment) Electrophoresis Blotting System
Humidity Range Up to 80%
View More Specs

Products 3
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
03-500-612
SDP
View Documents
Hoefer™
TE70XP
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (14 x 16cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
Each for $2,690.00
Add to cart
 
03-500-613
SDP
View Documents
Hoefer™
TE77XP
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
Each for $2,750.00
Add to cart
 
03-500-611
SDP
View Documents
Hoefer™
TE77X
Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ masks (16.5 x
Each for $2,130.00
Add to cart
 
Description

Description

  • Transfer up to two polyacrylamide gels in a stack
  • Use minimal buffer
  • Do not generate the excessive heat that can distort bands or denature protein
  • Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
  • Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
  • Available with built-in power supply to save space and simplify transfer

Certifications

CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1

Specifications

Specifications

500 mA
18.89 x 15 x 4 in. (48 x 38 x 10 cm)
Up to 80%
113°F
30 V
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Electrophoresis Blotting System
Semi Dry Transfer Unit
45°C
15 w
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit