Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Humboldt Hoffman Tubing Clamps
SDP
Regulate flow from fully open to closed
Manufacturer:  Humboldt H8740
 View more versions of this product
Catalog No. 05-870B
Add to cart
Description
Description
Regulate flow from fully open to closed
  • For flexible tubing
  • Nickel-plated brass
  • Won't rust
  • Lower jaw is either fixed or swivel-mounted for easy insertion of tubing without disconnecting apparatus
Specifications
Specifications
1 Ea.
Nickel Plated Brass
Control the flow in flexible tubing
0 to 0.75 in.
Fixed
Tubing Clamp
Control The Flow in Flexible Tubing
0 to 19 mm
1 in.
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Cancel Submit