Learn More
M+A Matting Hog Heaven Prime Mat
Andersen Hog Heaven Prime Mat is recommended for use in areas such as distribution, light manufacturing, work stations, check out stations, retail and healthcare environments
Manufacturer:  M+A Matting 4222060035
 View more versions of this product
Catalog No. 18-333-193
Add to cart
Description
Description
Features:
  • Urethane foam comfort
  • Nitrile rubber molded to a closed cell nitrile cushion for superior comfort and performance
  • Edge to edge cushion
  • High heel shoe safe, welding safe, and static dissipative
  • Anti-microbial treated for lifetime protection against odors and degradation
  • Beveled borders
  • Grease and oil proof, chemical resistant
Specifications
Specifications
Anti-microbial
3ft.
Chemical resistant mat
Beveled borders
5ft.