Learn More
Sklar InstrumentsSupplier Diversity Partner TC Cooley Microvascular Needle Holder, Sklar, 8 in., Straight, Smooth, Delicate, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 214951
TC Cooley Microvascular Needle Holder, Sklar, 8 in., Straight, Smooth, Delicate, Tungsten Carbide / Stainless Steel. Sklar Cooley Tungsten Carbide Microvascular Needle Holder is a ratcheted, finger ring instrument designed to hold small sutures. This needle holder is commonly used in microsurgery, plastic surgery, and cardiovascular procedures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is straight with smooth tips and a length of 8 inches. It features a very delicate pattern.
Catalog No. 50-143-7652