Learn More
EISCO Hollow Acrylic Semi Circle
The EISCO Hollow Acrylic Semi Circle is a handy devices for observing the index of refraction for different materials.
Manufacturer:  EISCO PH0585A
Catalog No. S12962
Add to cart
Description
Description
  • Half circle is made of acrylic with an indicator line half way across the arc
  • Dimensions are 3 in. x 1.5 in. x 0.75 in.
  • Typical uses for this device are reflection, refraction, wave demonstrations, Snell's law, observational study