Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Hollow Glass Prisms
Hollow glass prisms are made from glass plates that are cemented together. Stoppers included. Excellent tools for optical experimentation as well as for studying Snells Law of Refraction. Change the liquids or gases in the prisms to change the refraction properties
Supplier: Eisco™ PH0552C
Includes: prism with stopper
Description
- Made from glass plates, cemented together.
- Stopper included
- Hollow inside allows for a variety of optic experiments
- Excellent tool for studying Snells Law of Refraction.
- Designed for educational use
Warranty and Services
1 year limited manufacturer warranty
Specifications
Hollow Rectangular, Slab | |
Optics experiments, Snell's Law of Refraction | |
1 | |
150 g |
Glass | |
Prism, Stopper | |
8.9 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction