Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium Foil, 0.127mm (0.005 in.) thick, 99.9% (REO), Alfa Aesar™

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym
23988
CHEBI:49648
holmium
[Ho]
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym
23988
CHEBI:49648
holmium
[Ho]

Form: Approx.0.70g/25 x 25mm

Assay Percent Range 99.9% (REO)
Appearance Silver gray foil
Physical Form Foil, 0.127mm (0.005 in.) thick
Odor Odorless
Color Silver to Gray
Quantity 50 x 50mm
Chemical Name or Material Holmium

EINECSNumber : 231-169-0

TSCA : Yes

Recommended Storage : Ambient temperatures