Alfa Aesar™ Holmium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1319909

Quantity 10g

 View more versions of this product

Catalog No. AA1319909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14589-33-4
    Formula Weight 462.26
    MDL Number MFCD00050176
    Synonym Holmium acetylacetonate
    Chemical Name or Material Holmium(III) 2,4-pentanedionate
    Grade Reacton™

    Specifications

    Formula Weight 462.26
    Odor Odorless
    Physical Form Powder
    Sensitivity Hygroscopic
    MDL Number MFCD00050176
    Quantity 10g
    CAS 14589-33-4
    Synonym Holmium acetylacetonate
    Chemical Name or Material Holmium(III) 2,4-pentanedionate
    Assay 99.9%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit