Alfa Aesar™ Holmium(III) isopropoxide, 5% w/v in toluene/isopropanol

Manufacturer: Alfa Aesar™ 36579AC

Quantity 25mL

 View more versions of this product

Catalog No. AA36579AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 30429-72-2
    Linear Formula Ho[OCH(CH3)2]3
    Formula Weight 342.19
    MDL Number MFCD00144455
    UN Number UN1993
    Chemical Name or Material Holmium(III) isopropoxide

    Specifications

    Formula Weight 342.19
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Pale red
    Physical Form Liquid
    Linear Formula Ho[OCH(CH3)2]3
    UN Number UN1993
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Sensitivity Moisture Sensitive
    Quantity 25mL
    MDL Number MFCD00144455
    CAS 30429-72-2
    Concentration 5% w/v in toluene/isopropanol
    Chemical Name or Material Holmium(III) isopropoxide

    GHS H Statement
    H225-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit