Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
12055-62-8
Ho2O3
377.85
MFCD00011053
12055-62-8
Ho2O3
377.85
MFCD00011053
for ultra trace analysis
Assay Percent Range 99.85% min. (Ho2O3) TREO base
Linear Formula HO2O3
Merck Index 15, 4765
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inorganic acids
Formula Weight 377.85
Physical Form Powder
Percent Purity 99.9%
Grade (Total Rare Earth Oxides)
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in inorganic acids
Boiling Point 3900.0°C
Color Beige to Orange-Yellow
Melting Point 2330.0°C
Quantity 10g
Chemical Name or Material Holmium(III) oxide
Show More Show Less

EINECSNumber : 235-015-3