Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Holmium(III) sulfate hydrate, 99.99%

Click to view available options
Quantity:
5g
25g
15622-40-9
Ho2O12S3·xH2O
MFCD00149740
15622-40-9
Ho2O12S3·xH2O
MFCD00149740
Linear Formula Ho2(SO4)3·xH2O
Formula Weight 618.03 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Quantity 5g
Chemical Name or Material Holmium(III) sulfate hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 239-697-3

TSCA : Yes

Recommended Storage : Ambient temperatures