Alfa Aesar™ Holmium rod, 6.35mm (0.25 in.) dia., 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 44050HF

Quantity 10mm

 View more versions of this product

Catalog No. AA44050HF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-60-0
    MDL Number MFCD00011049
    Chemical Name or Material Holmium rod, ≈6.35mm (0.25 in.) dia.

    Specifications

    Physical Form Cast Rod
    Quantity 10mm
    Packaging Mylar under Argon
    MDL Number MFCD00011049
    Odor Odorless
    CAS 7440-60-0
    Weight ≈0.28g/mm
    TSCA TSCA
    Storage Note 1
    • Packaged in mylar under argon
    Chemical Name or Material Holmium rod, ≈6.35mm (0.25 in.) dia.
    Assay 99.9%, (metals basis)

    • Packaged in mylar under argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit