Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMER2 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271464AF647

Catalog No. N271464A647


Add to cart

Description

Description

HOMER2 Monoclonal antibody specifically detects HOMER2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HOMER2
OTI1D1
Western Blot
Affinity Purified, Conjugated
HOMER2
Full length recombinant protein of human HOMER2 (NP_004830) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ACPD, CPD, cupidin, homer homolog 2 (Drosophila), homer homolog 3, homer protein homolog 2, homer, neuronal immediate early gene, 2, Homer-2, HOMER2A, HOMER-2A, HOMER2B, HOMER-2B, Vesl-2
Mouse
IgG2b
0.1 mL
Primary
9455.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit