Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homing Receptor Mouse anti-Guinea Pig, Clone: CT4, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Homing Receptor
Clone CT4
Dilution Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:100-1:1000
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10064936
View Documents
Novus Biologicals
NB10064936
0.125 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Homing Receptor Monoclonal specifically detects Homing Receptor in Guinea Pig samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Homing Receptor
Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:100-1:1000
Monoclonal
Ascites
RUO
Ascites with 0.09% Sodium Azide
IgG3
Unpurified
Antibody recognizes guinea pig THY1 (CD90) a small but heavily glycosylated member of the immunoglobulin superfamily. Clone CT4 was originally identified as recognizing a guinea pig homing receptor (Kraal et al. 1986) and later confirmed as recognizing the guinea pig homologue of human and rodent CD90 (Schafer et al. 1999) by purification and microsequencing. Guinea Pig CD90 is notable for its level of Nlinked glycosylation, rendering it an apparent molecular mass of ∼36kDa, higher than that seen in most other species where CD90 has been identified and characterized.
CT4
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Guinea Pig
Peritoneal T-cells
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit