Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homophthalic anhydride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
703-59-3
C9H6O3
162.14
MFCD00006894
AKHSBAVQPIRVAG-UHFFFAOYSA-N
homophthalic anhydride,1,3-isochromandione,isochroman-1,3-dione,1h-2-benzopyran-1,3 4h-dione,homophthalic acid anhydride,4h-2-benzopyran-1,3-dione,1h-isochromene-1,3 4h-dione,3,4-dihydro-1h-2-benzopyran-1,3-dione,4h-benzo c pyran-1,3-dione,homophthalic anhydride 2-carboxyphenylacetic anhydride
12801
4H-isochromene-1,3-dione
C1C2=CC=CC=C2C(=O)OC1=O
703-59-3
C9H6O3
162.14
MFCD00006894
AKHSBAVQPIRVAG-UHFFFAOYSA-N
homophthalic anhydride,1,3-isochromandione,isochroman-1,3-dione,1h-2-benzopyran-1,3 4h-dione,homophthalic acid anhydride,4h-2-benzopyran-1,3-dione,1h-isochromene-1,3 4h-dione,3,4-dihydro-1h-2-benzopyran-1,3-dione,4h-benzo c pyran-1,3-dione,homophthalic anhydride 2-carboxyphenylacetic anhydride
12801
4H-isochromene-1,3-dione
C1C2=CC=CC=C2C(=O)OC1=O
Melting Point 140.0°C to 144.0°C
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
Color Yellow to Beige
Assay Percent Range 98%
Beilstein 17, 489
Fieser 11,254
Formula Weight 162.14
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Homophthalic anhydride
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-873-4