Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homopiperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
505-66-8
C5H12N2
100.165
MFCD00006933
FQUYSHZXSKYCSY-UHFFFAOYSA-N
homopiperazine,1,4-diazacycloheptane,1h-1,4-diazepine, hexahydro,hexahydro-1,4-diazepine,1,4 diazepane,perhydro-1,4-diazepine,trimethyleneethylenediamine,1,4-diazepan,unii-95cl167w8t,hexahydro-1h-1,4-diazepine
68163
1,4-diazepane
C1CNCCNC1
505-66-8
C5H12N2
100.165
MFCD00006933
FQUYSHZXSKYCSY-UHFFFAOYSA-N
homopiperazine,1,4-diazacycloheptane,1h-1,4-diazepine, hexahydro,hexahydro-1,4-diazepine,1,4 diazepane,perhydro-1,4-diazepine,trimethyleneethylenediamine,1,4-diazepan,unii-95cl167w8t,hexahydro-1h-1,4-diazepine
68163
1,4-diazepane
C1CNCCNC1
Water 1% max.
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 64°C
Color White to Yellow
Melting Point 42.0°C to 45.0°C
Quantity 500g
Assay Percent Range 98%
Formula Weight 100.16
Physical Form Crystalline Mass
Percent Purity 98%
Chemical Name or Material Homopiperazine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger