Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homoquinolinic acid, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

490-75-5
C8H7NO4
181.147
HQPMJFFEXJELOQ-UHFFFAOYSA-N
3080554

Selective, potent NMDA agonist

Potent NMDA receptor agonist; subtype-selective.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
CAS 490-75-5
Molecular Formula C8H7NO4
InChI Key HQPMJFFEXJELOQ-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)C(=O)O)CC(=O)O
IUPAC Name 3-(carboxymethyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol) 181.147
PubChem CID 3080554
Formula Weight 181.15
Chemical Name or Material Homoquinolinic acid
Grade >99%
Show More Show Less

Recommended Storage : Store at Room Temperature