Learn More
Homosalate, Spectrum™
Click to view available options
Quantity:
25 g
100 g
1 kg
20 kg
200 kg
Packaging:
Amber Glass Bottle
Drum
Poly Drum
118-56-9
C16H22O3
262.35
WSSJONWNBBTCMG-UHFFFAOYNA-N
3,3,5-trimethylcyclohexyl 2-hydroxybenzoate
CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1O
118-56-9
C16H22O3
262.35
WSSJONWNBBTCMG-UHFFFAOYNA-N
3,3,5-trimethylcyclohexyl 2-hydroxybenzoate
CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1O
H1187, 118-56-9, C16H22O3
Spectrum™ Homosalate is an organic compound most often used in sunscreens as a chemical UV filter. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 1