Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homosalate, USP, 90-110%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
20 kg
200 kg
Packaging:
Amber Glass Bottle
Fiber Drum
118-56-9
C16H22O3
262.35
WSSJONWNBBTCMG-UHFFFAOYNA-N
3,3,5-trimethylcyclohexyl 2-hydroxybenzoate
CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1O
118-56-9
C16H22O3
262.35
WSSJONWNBBTCMG-UHFFFAOYNA-N
3,3,5-trimethylcyclohexyl 2-hydroxybenzoate
CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1O

H1121, 118-56-9, C16H22O3

Spectrum™ Homosalate, USP is a common sunscreen ingredient as a chemical UV filter. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Refractive Index 1.516-1.519
Quantity 1 kg
Specific Gravity 1.049-1.053
CAS Max % 1
Percent Purity 90 to 110%