Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homovanillic acid, Tocris Bioscience™

Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
306-08-1
C9H10O4
182.175
QRMZSPFSDQBLIX-UHFFFAOYSA-N
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
1738
CHEBI:545959
2-(4-hydroxy-3-methoxyphenyl)acetic acid
COC1=C(C=CC(=C1)CC(=O)O)O
This item is not returnable. View return policy
306-08-1
C9H10O4
182.175
QRMZSPFSDQBLIX-UHFFFAOYSA-N
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
1738
CHEBI:545959
2-(4-hydroxy-3-methoxyphenyl)acetic acid
COC1=C(C=CC(=C1)CC(=O)O)O

Fluorimetric reagent

Fluorimetric reagent and major catecholamine metabolite. Used for fluorimetric determination of oxidative enzymes including glucose oxidase.
Formula Weight 182.17
Percent Purity >99%
Quantity 50 mg
Chemical Name or Material Homovanillic acid

Recommended Storage : Store at +4°C