Learn More
Tele-Lite™ Honda™ Lightweight Pumps
Manufacturer:  Tele-Lite™ WX10K1AT
 View more versions of this product
Catalog No. 19-120-2689
Add to cart
Specifications
Specifications
51psi
13.4 lb.
Single cylinder mini-engine, air cooled, GX25, self priming, suction and discharge port
37 gal./min.
29.97cm
32.51cm
General purpose, centrifugal, self priming
1.5hp
6.07kg
WX10
11.8 in.
12.8 in.
8.7 in.