Learn More
Tele-Lite™ Honda™ Portable Economy Generators
Manufacturer:  Tele-Lite™ EG5000CLAT
Catalog No. 19-120-2674
Add to cart
Specifications
Specifications
41.2/20.8A
120/240V
23.84L
389cc
Refrigerator or freezer (energy star), microwave oven, TV/DVD, hair dryer, coffee maker, incandescent lights, air compressor, bench grinder (8 in.), circular saw (heavy duty, 7.25 in.), concrete vibrator, drills, high-pressure washer (1hp), table saw (10
22.6 in.
Electronic
26.8 in.
68cm
Locking plug
Recoil
22.8 in.
57.91cm
5000w
26.8 x 22.8 x 22.6 in. (68 x 57.91 x 57.4cm)
60Hz
6.3 gal.
Air cooled
Honda GX390 4-stroke overhead valve, single cylinder
Protects the generator by shutting the engine off when low oil is detected and also from being overloaded
11hp
57.4cm
Voltage selector switch, tough one-piece welded frame
73dB
11 Hours at 0.5 load
177 lb.
80.28kg
17060.7 BTU/h
6.3 gal., 23.84L
120/240V 60Hz