Honeywell Analytics™ Biosystems Multi-Component Calibration Gas Mixtures

Manufacturer: Honeywell Analytics™ 549043

 View more versions of this product

Catalog No. 19-088-615

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 103L
    Product Type Mulit component gas calibration cylinder
    For Use With (Equipment) Biosystem
    Includes 50% LEL Methane, 50ppm Carbon Monoxide, 25ppm Hydrogen Sulphide, 18% Oxygen, Nitrogen balance Gas
    Shelf Life 5 Years
    Capacity 3482.8 oz., 103L
    Capacity (English) 3482.8 oz.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit