Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors

For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.

Manufacturer: Honeywell Analytics™ GAXTXDL2

 View more versions of this product

Catalog No. 19413413

  Check Availability
Add to Cart

    Specifications

    Product Type Oxygen Detector
    Warranty 2 Years
    Certifications/Compliance ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC
    Depth (English) 1.1 in.
    Depth (Metric) 2.8cm
    Height (English) 3.75 in.
    Height (Metric) 9.52cm
    Humidity Range 0 to 99%
    Measurement Range 0 to 30%
    Sensor Types Oxygen
    Weight (English) 2.9 oz.
    Weight (Metric) 83g
    Width (English) 2 in.
    Width (Metric) 5.08cm
    Temperature (English) Range -4° to +122°F
    Temperature (Metric) Range -20° to +50°C

    • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
    • Secure and accurate history with datalogging with passcode protection
    • Continuous LCD shows real-time gas concentrations
    • Simple automatic calibration procedure
    • Equipped with internal vibrating alarm for high noise areas
    • Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
    • Replaceable 3V battery
    • Water-resistant
    • Bright wide-angled visual alarm bars
    • Two-year field-replaceable battery (1.5 year life) and sensor
    • Adjustable alarm setpoints
    • Multilanguage display options 
    • Compatible with Sampler motorized pump
    • Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
    • Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
    • Size: 2.8 x 5.0 x 9.5cm (1.1 x 2.0 x 3.75 in.)
    • Weight: 82g (2.9 oz.)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit