Learn More
Honeywell Analytics™ GasAlertClip Extreme Single-Gas Detectors
Durable, reliable, accurate, easy to use
Manufacturer:  Honeywell Analytics™ GA24XTH
 View more versions of this product
Catalog No. 19-807-965
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
5/10 or 10/15ppm
-40° to +50°C
Visual, vibrating, audible (95 dB)
ATEX: CE III 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, IECEx: Exia IIC T4, INMETRO: BR Exia IIC T4, EMC Directive 89/336/EEC
2.79cm
3.2 in.
5 to 95%
5.08cm
Hydrogen Sulfide
76g
5.08cm
-40° to +122°F
2 Years
2 Years
1.1 in.
LCD
8.12cm
66/67
0 to 100ppm
2.7 oz.
2 in.