Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ GasAlertMax XT II Multigas Detectors

Monitor up to four hazardous gases with a robust, motorized pump for intelligent, remote sampling. The GasAlertMax XT II Multigas Detector is compact, comfortable, field tough, and ideal for confined sampling.

Manufacturer:  Honeywell Analytics™ XTX00MYNA

 View more versions of this product

Catalog No. 19-413-380


  Check Availability
Add to cart

Specifications

Description

  • Integrated sampling pump with reliable diaphragm technology
  • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
  • Equipped with standard datalogging and event logging
  • Continuous LCD shows real-time gas concentrations
  • Block detection with semi-conductor pressure sensor
  • User adjustable alarm setpoints
  • Multi-language support
  • Water-resistant
  • Size: 13.1 x 7.0 x 5.2cm (5.1 x 2.8 x 2.0 in.)
  • Weight: 316g (11.1 oz.)
  • Temperature Range: -20° to +50°C (-4° to +122°F)
  • Humidity: 10 to 100% RH (non-condensing)

 

H2S Sensors

  • Measuring Range: 0 to 200ppm
  • Resolution: 1.0ppm
CO Sensors
  • Measuring Range: 0 to 1000ppm
  • Resolution: 1.0ppm
O2 Sensors
  • Measuring Range: 0 to 30% by vol
  • Resolution: 0.1%
Combustible Gas Sensors
  • Measuring Range: 0 to 100% LEL; 0 to 5.0% v/v
  • Resolution: 1.0% LEL; 0.1% v/v
Description & Specifications

Specifications

Full 2 year warranty including all sensors
8 to 13 hr.
2 in.
LCD
13cm
O2: 0.1%, CO: 1ppm
-4° to +122°F
11.1 oz.
2.8 in.
Audible, visual, vibrating
Yellow
5.08cm
5.12 in.
10 to 100%
Carbon Monoxide, Oxygen
-20° to +50°C
316g
7.2cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit