Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ GasAlertQuattro Multigas Detectors

Manufacturer:  Honeywell Analytics™ QTXWH0ABNA

 View more versions of this product

Catalog No. 19413333


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Audible, visual, vibrating
Alkaline
1.87 in.
5.12 in.
10 to 100%
Hydrogen Sulfide, LEL, Oxygen
-20° to +50°C
338g
8.07cm
14 hr.
Black
4.7cm
13cm
O2: 0.1%, LEL: 1%, H2S: 1ppm
-4° to +122°F
11.92 oz.
3.18 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit