Learn More
Honeywell Analytics™ GasAlertQuattro Rechargable Multigas Detectors
Supplier:  Honeywell Analytics™ QTX0H0RYNA
 View more versions of this product
Catalog No. 19-413-324
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Audible, visual, vibrating
Yellow
4.7 cm
5.12 in.
10 to 100%
-4°F to +122°F
11.15 oz.
3.25 in.
18 to 20 hr.
1.87 in.
Hydrogen Sulfide, Oxygen
13 cm
O2: 0.1%, H2S: 1ppm
-20°C to +50°C
316g
8.25 cm
Product Suggestions
Product Suggestions