Learn More
Honeywell Analytics™ IAQPoint2 CO2 Monitors
Manufacturer:  Honeywell Analytics™ 1508A2001
 View more versions of this product
Catalog No. 19-413-458
  Check Availability
Add to cart
Specifications
Description
  • Fully functional, multi-color, touch screen interface
  • Operates on +24 VAC/VDC
  • Output: 4 to 20mA or 2 to 10 VDC
  • Switch selectable
  • Duct mount configuration
  • Low power consumption and ‘sleep mode’ feature
User‑Friendly Touchscreen Operation
  • Menu driven, customizable, easy-to-navigate display
  • Large 4.9 x 6.5cm (1.9 x 2.6 in.) VGA screen with easy-to-read LCD digits and graphical display
Applications
Auditoriums, Conference rooms, Schools, Hospitals, Washrooms, Kitchens, Janitorial closets, Copier/printing rooms, Warehouses/shipping areas
Certifications
Intertek Listed to 3rd Editions, Intertek tested to EN. LonWorks
Compliance
Meets ASHRAE 15 standards
Warranty
5 year sensor life with automatic self-calibration
Description & Specifications
Specifications
VGA screen with easy-to-read LCD digits and graphical display
Iaqpoint2 Carbon Dioxide Monitor
CO2
Meets ASHRAE 15 standards; Intertek Listed to 3rd Editions, Intertek tested to EN. LonWorks
4 to 20mA or 2 to 10 VDC
+24 VAC/VDC
Analog